BioLite™ Microwell Plates
Thermo Scientific™
BioLite™ Microwell Plates
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Plates that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsCulture AreaVolume (Metric) Well
130188960.32 cm20.2 mL
13018469.6 cm23 mL
130185123.8 cm22 mL
130186241.9 cm21 mL
130187480.75 cm20.5 mL
Catalog number 130188
Price (MXN)
-
No. of Wells:
96
Culture Area:
0.32 cm2
Volume (Metric) Well:
0.2 mL
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated plates with lids suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Sterile per ISO 11137, SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• BSE/TSE Free
• RNase/DNase Free
• USP Class VI raw material
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Culture Area0.32 cm2
FormatStandard
LidWith Lid
No. per Case50
No. per Pack1/Pk.
Packaging1/Pk., 50/Cs.
Product TypeMicroplate
Shelf Life2 years
Volume (Metric) Well0.2 mL
Volume (Metric) Working0.2 mL
Array8 x 12
ColorClear
No. of Wells96
Product LineBiolite
SterilitySterile
Surface TreatmentCell Culture
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50
Frequently asked questions (FAQs)
What is the recommended working volume of Nunc adherent cell culture products?
This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.