4-Phenoxyaniline, 97%
4-Phenoxyaniline, 97%
4-Phenoxyaniline, 97%
Thermo Scientific Chemicals

4-Phenoxyaniline, 97%

CAS: 139-59-3 | C12H11NO | 185.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 130235000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS139-59-3
IUPAC Name4-phenoxyaniline
Molecular FormulaC12H11NO
InChI KeyWOYZXEVUWXQVNV-UHFFFAOYSA-N
SMILESNC1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes
Melting point82°C to 86°C
Appearance (Color)Brown to green-brown
Infrared spectrumConforms
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only