Interested in a bulk or custom version of this product? Request pricing.

IGF2 Recombinant Human Protein (His.NusA Tag)

Catalog number: 13032H24E25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Insulin-like growth factor 2 (IGF2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF2 (P01344-1) (Ala 25-Glu 91) fused to the polyhistidine-NusA tag at the N-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human IGF2 at 10 µg/ml(100 µl/well) can bind biotinylated human His-IGFBP3. The EC50 of binding to biotinylated human His-IGFBP3 is 18 ng/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Development, Neurobiology, Other Research
Expression: E. coli
Form: Lyophilized
Gene Aliases: PP1446, C11orf43, FLJ22066, FLJ44734, IGF-II, PP9974
Gene Symbol: IGF2
Molecular Weight: 64.6 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Growth Factors & Receptors
Protein Form: Full Length, Recombinant
Protein Name: insulin-like growth factor 2 (somatomedin A)
Protein Subtype: IGFs (Insulin-Like Growth Factors)
Protein Tag or Fusion: His Tag, NusA Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products