4-Phenylbutyric acid, 99%
4-Phenylbutyric acid, 99%
4-Phenylbutyric acid, 99%
Thermo Scientific Chemicals

4-Phenylbutyric acid, 99%

CAS: 1821-12-1 | C10H12O2 | 164.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 130381000
Price (EUR)
121,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
121,00
Each
Chemical Identifiers
CAS1821-12-1
IUPAC Name4-phenylbutanoic acid
Molecular FormulaC10H12O2
InChI KeyOBKXEAXTFZPCHS-UHFFFAOYSA-N
SMILESOC(=O)CCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH98.5 to 101.5 %
Appearance (Form)Powder or crystals and/or chunks or fused solid
Appearance (Color)White to light yellow to beige
Melting point49°C to 53°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only