1,3-Phenylenediacetic acid, 97%
1,3-Phenylenediacetic acid, 97%
1,3-Phenylenediacetic acid, 97%
Thermo Scientific Chemicals

1,3-Phenylenediacetic acid, 97%

CAS: 19806-17-8 | C10H10O4 | 194.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1305000101 gGlass bottle
Catalog number 130500010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range97%
Beilstein09, 874
CAS Max %100.0
CAS Min %96.0
CAS19806-17-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only