Search
| Número do catálogo | Includes |
|---|---|
| 1305600 | Injector 2.5 mm ID quartz |
| BRE0017466 | Injector 2.0 mm, qz, 1-inlet |
| 1305640 | Injector 2.0 mm ID quartz |
| 1321200 | Injector 2.0 mm ID platinum |
| 1323600 | Injector 2.0 mm ID sapphire |
| 1323630 | Injector 2.0 mm, qz, 2- inlet |
| 1325110 | Injector 1.0 mm ID quartz |
| 1305650 | Injector 1.5 mm ID quartz |
Choose from this range of reliable, screw-in, self-aligning injectors, with differing internal diameters and materials, to match your sample matrices and analytical needs.
The iCAP MX and iCAP Q/Qnova Series ICP-MS systems come as standard with a 2.5 mm internal diameter, quartz injector. All of our injectors are easily removable for improved robustness and reduced maintenance requirements.
Choose high-quality consumables to improve performance