m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
Thermo Scientific Chemicals

m-Phenylenediamine, 99+%

CAS: 108-45-2 | C6H8N2 | 108.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
檢視全部
Glass bottle
產品號碼 130560010
價格 (TWD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS108-45-2
IUPAC Namebenzene-1,3-diamine
Molecular FormulaC6H8N2
InChI KeyWZCQRUWWHSTZEM-UHFFFAOYSA-N
SMILESNC1=CC(N)=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Flakes or pellets
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)White to tan to grey
AppearanceMay darken on storage
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only