Thermo Scientific Chemicals

m-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Catalog number: 130560250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

m-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Catalog number: 130560250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130560250
Price (USD)
Price: 27.70
Online price: 23.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-45-2
IUPAC Name
benzene-1,3-diamine
Molecular Formula
C6H8N2
InChI Key
WZCQRUWWHSTZEM-UHFFFAOYSA-N
SMILES
NC1=CC(N)=CC=C1
Appearance
May darken on storage
Melting point
62°C to 65°C
Appearance (Color)
White to tan to grey
Appearance (Form)
Flakes or pellets
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text