m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
m-Phenylenediamine, 99+%
Thermo Scientific Chemicals

m-Phenylenediamine, 99+%

CAS: 108-45-2 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
500 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 130562500
Price (EUR)
28,65
Online exclusive
32,10
Save 3,45 (11%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
28,65
Online exclusive
32,10
Save 3,45 (11%)
Each
Chemical Identifiers
CAS108-45-2
IUPAC Namebenzene-1,3-diamine
Molecular FormulaC6H8N2
InChI KeyWZCQRUWWHSTZEM-UHFFFAOYSA-N
SMILESNC1=CC(N)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan to grey
AppearanceMay darken on storage
Melting point62°C to 65°C
Appearance (Form)Flakes or pellets
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only