p-Phenylenediamine, 99+%
p-Phenylenediamine, 99+%
p-Phenylenediamine, 99+%
Thermo Scientific Chemicals

p-Phenylenediamine, 99+%

CAS: 106-50-3 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 130575000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-50-3
IUPAC Namebenzene-1,4-diamine
Molecular FormulaC6H8N2
InChI KeyCBCKQZAAMUWICA-UHFFFAOYSA-N
SMILESNC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes
Infrared spectrumConforms
AppearanceMay darken on storage
GC>=99.0 %
Appearance (Color)White to grey to brown to purple
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only