Phenyl ether, 99%
Phenyl ether, 99%
Phenyl ether, 99%
Thermo Scientific Chemicals

Phenyl ether, 99%

CAS: 101-84-8 | C12H10O | 170.21 g/mol
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 130600025
Price (JPY)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS101-84-8
IUPAC Namephenoxybenzene
Molecular FormulaC12H10O
InChI KeyUSIUVYZYUHIAEV-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid after melting
Infrared spectrumConforms
Refractive index1.5790 to 1.5820 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only