DL-alpha-Phenylglycine, 99%
DL-alpha-Phenylglycine, 99%
DL-alpha-Phenylglycine, 99%
DL-alpha-Phenylglycine, 99%
Thermo Scientific Chemicals

DL-alpha-Phenylglycine, 99%

CAS: 2835-06-5 | C8H9NO2 | 151.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
130641000100 gPlastic bottle
Catalog number 130641000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2835-06-5
IUPAC Name2-amino-2-phenylacetic acid
Molecular FormulaC8H9NO2
InChI KeyZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILESNC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder
Appearance of solution(5% in 1N HCl) clear colorless to slightly yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only