N-Phenylmaleimide, 97%
N-Phenylmaleimide, 97%
N-Phenylmaleimide, 97%
Thermo Scientific Chemicals

N-Phenylmaleimide, 97%

CAS: 941-69-5 | C10H7NO2 | 173.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
130711000100 gGlass bottle
Catalog number 130711000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS941-69-5
IUPAC Name1-phenyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular FormulaC10H7NO2
InChI KeyHIDBROSJWZYGSZ-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)N1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Melting point86°C to 91°C
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only