Phenylpropiolic acid, 97%
Phenylpropiolic acid, 97%
Phenylpropiolic acid, 97%
Thermo Scientific Chemicals

Phenylpropiolic acid, 97%

CAS: 637-44-5 | C9H6O2 | 146.15 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 130850050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS637-44-5
IUPAC Name3-phenylprop-2-ynoic acid
Molecular FormulaC9H6O2
InChI KeyXNERWVPQCYSMLC-UHFFFAOYSA-N
SMILESOC(=O)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=96.0 %
Melting point136°C to 139°C
Infrared spectrumConforms
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only