Regulatory requirements vary by country; products may not be available in your geographic area.
Plastic Sampling Spoon for Cattle
Catalog number: 13100
 Related applications: Animal Health
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This is a plastic spoon developed specifically for the sampling of bovine brain tissue. The spoon was designed to meet the requirements of slaughterhouse operation, and features sharp edges, allowing easy and safe removal of brain tissue through the spinal cord canal. The special design and the long handle prevent the staff from coming into contact with animal tissue during sampling.
For Veterinary Use Only.
Specifications
Product Line: Prionics™
Product Size: 1 piece
Type: Sample Collection Accessory
Documents
Manuals & protocols