Phloroglucinol Dihydrate, 99%
Phloroglucinol Dihydrate, 99%
Phloroglucinol Dihydrate, 99%
Thermo Scientific Chemicals

Phloroglucinol Dihydrate, 99%

CAS: 6099-90-7 | C6H6O3·2H2O | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 131041000
Price (EUR)
81,30
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
81,30
Each
Chemical Identifiers
CAS6099-90-7
IUPAC Namebenzene-1,3,5-triol dihydrate
Molecular FormulaC6H10O5
InChI KeyMPYXTIHPALVENR-UHFFFAOYSA-N
SMILESO.O.OC1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to beige
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only