Phthalide, 99%
Phthalide, 99%
Phthalide, 99%
Thermo Scientific Chemicals

Phthalide, 99%

CAS: 87-41-2 | C8H6O2 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 131095000
Price (USD)
129.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
129.00
Each
Chemical Identifiers
CAS87-41-2
IUPAC Name1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC8H6O2
InChI KeyWNZQDUSMALZDQF-UHFFFAOYSA-N
SMILESO=C1OCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
GC>=98.5 %
Melting point70°C to 73°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only