Thermo Scientific™
Shandon™ M-1 Embedding Matrix
Related applications:
Anatomical Pathology
Obtain optimal sample preparation with Thermo Scientific™ Shandon™ M-1 embedding matrix.
Back to top
  • Solution is clear and water-soluable
  • Optimum sectioning temperature of -12° to -13°C (-10.4° to -8.6°F)
  • Particularly effective on specimens with high water content such as eyes and CNS