Phthalimide, 99%
Phthalimide, 99%
Phthalimide, 99%
Thermo Scientific Chemicals

Phthalimide, 99%

CAS: 85-41-6 | C8H5NO2 | 147.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
產品號碼 131100025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS85-41-6
IUPAC Name2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC8H5NO2
InChI KeyXKJCHHZQLQNZHY-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CC=CC=C12
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder or flakes
Infrared spectrumConforms
Melting point232°C to 236°C
HPLC>=98.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only