Pimelic Acid, 98%
Pimelic Acid, 98%
Pimelic Acid, 98%
Acros Organics

Pimelic Acid, 98%

CAS: 111-16-0 | C7H12O4 | 160.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
131231000
also known as 13123
100 gGlass bottle
Catalog number 131231000
also known as 13123
HSN 29171990
Price (INR)
17,786.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
17,786.00
Each
Chemical Identifiers
CAS111-16-0
IUPAC Nameheptanedioic acid
Molecular FormulaC7H12O4
InChI KeyWLJVNTCWHIRURA-UHFFFAOYSA-N
SMILESOC(=O)CCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point103°C to 106°C
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only