1,3-Propanedithiol, 98%
1,3-Propanedithiol, 98%
1,3-Propanedithiol, 98%
Thermo Scientific Chemicals

1,3-Propanedithiol, 98%

CAS: 109-80-8 | C3H8S2 | 108.22 g/mol
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 131412500
Price (JPY)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-80-8
IUPAC Namepropane-1,3-dithiol
Molecular FormulaC3H8S2
InChI KeyZJLMKPKYJBQJNH-UHFFFAOYSA-N
SMILESSCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5380 to 1.5420 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only