2-Propanethiol, 98%
2-Propanethiol, 98%
2-Propanethiol, 98%
Thermo Scientific Chemicals

2-Propanethiol, 98%

CAS: 75-33-2 | C3H8S | 76.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 131445000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-33-2
IUPAC Namepropane-2-thiol
Molecular FormulaC3H8S
InChI KeyKJRCEJOSASVSRA-UHFFFAOYSA-N
SMILESCC(C)S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4240 to 1.4270 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only