Propargylamine hydrochloride, 95%
Propargylamine hydrochloride, 95%
Propargylamine hydrochloride, 95%
Thermo Scientific Chemicals

Propargylamine hydrochloride, 95%

CAS: 15430-52-1 | C3H6ClN | 91.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 131470010
Price (EUR)
36,65
Online exclusive
40,60
Save 3,95 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
36,65
Online exclusive
40,60
Save 3,95 (10%)
Each
Chemical Identifiers
CAS15430-52-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to brown
Appearance (Form)Powder or crystals or crystalline powder
Infrared spectrumConforms
Melting point174°C to 184°C
Titration Argentometric94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only