Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution
Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution
Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution
Gibco™

Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution

Gibco™ Cell Dissociation Buffer는 무칼슘, 무마그네슘 Hank's Balanced Salt Solution (HBSS)에 염, 킬레이트제, 세포조절제가 들어있는 막여과된 등장성 무효소 용액입니다. Gibco™자세히 알아보기
Have Questions?
카탈로그 번호수량
13150016100 mL
카탈로그 번호 13150016
제품 가격(KRW)
78,000
온라인 행사
Ends: 31-Dec-2026
86,000
할인액 8,000 (9%)
Each
수량:
100 mL
제품 가격(KRW)
78,000
온라인 행사
Ends: 31-Dec-2026
86,000
할인액 8,000 (9%)
Each
Gibco™ Cell Dissociation Buffer는 무칼슘, 무마그네슘 Hank's Balanced Salt Solution (HBSS)에 염, 킬레이트제, 세포조절제가 들어있는 막여과된 등장성 무효소 용액입니다. Gibco™ Cell Dissociation Buffer는 리간드 결합, 유세포분석, 면역조직화학 등 세포 표면이 온전한 단백질이 필요한 연구에서 포유류 세포를 무리없이 해리하는데 적합합니다. Gibco™ Cell Dissociation Buffer는 HeLa 및 NIH 3T3 등 약하게 붙어있는 세포주에 적합하며 강하게 부착된 세포의 일반적 계대배양에 권장되지 않습니다. dissociation protocol을 본사 Technical Reference Library에서 이용할 수 있습니다.
For Research Use Only. Not for use in diagnostic procedures.
사양
세포주HeLa, NIH 3T3
킬레이터EDTA
수량100 mL
비고Room Temperature Stable
유통 기한24 Months
배송 조건Room Temperature
분류Animal Origin-free
제품 유형Cell Culture Dissociation Reagent
멸균Sterile-filtered
첨가제 포함EDTA, Earle's Balanced Salt Solution (EBSS)
Unit SizeEach
구성 및 보관
Storage conditions: 15–30°C
Shipping conditions: ambient
Shelf life: 24 months from date of manufacture

자주 묻는 질문(FAQ)

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
The Low Density Lipoprotein Receptor-related Protein Mediates Fibronectin Catabolism and Inhibits Fibronectin Accumulation on Cell Surfaces.
Authors: Salicioni Ana M; Mizelle Kellie S; Loukinova Elena; Mikhailenko Irina; Strickland Dudley K; Gonias Steven L;
Journal:J Biol Chem
PubMed ID:11867643
Low density lipoprotein receptor-related protein (LRP) is a member of the low density lipoprotein receptor family, which functions as an endocytic receptor for diverse ligands. In this study, we demonstrate that murine embryonic fibroblasts (MEF-2 cells) and 13-5-1 Chinese hamster ovary cells, which are LRP-deficient, accumulate greatly increased levels of ... More