Propyl gallate, 98%
Propyl gallate, 98%
Propyl gallate, 98%
Thermo Scientific Chemicals

Propyl gallate, 98%

CAS: 121-79-9 | C10H12O5 | 212.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 131585000
Price (USD)
253.65
Online exclusive
282.00
Save 28.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
253.65
Online exclusive
282.00
Save 28.35
Each
Chemical Identifiers
CAS121-79-9
IUPAC Namepropyl 3,4,5-trihydroxybenzoate
Molecular FormulaC10H12O5
InChI KeyZTHYODDOHIVTJV-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (110°C, 4 hrs)
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only