4-n-Propylphenol, 99%
4-n-Propylphenol, 99%
4-n-Propylphenol, 99%
Thermo Scientific Chemicals

4-n-Propylphenol, 99%

CAS: 645-56-7 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 131620050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS645-56-7
IUPAC Name4-propylphenol
Molecular FormulaC9H12O
InChI KeyKLSLBUSXWBJMEC-UHFFFAOYSA-N
SMILESCCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only