Pyrazine, 99+%
Pyrazine, 99+%
Pyrazine, 99+%
Acros Organics

Pyrazine, 99+%

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
131700250
also known as 13170
25 gGlass bottle
Catalog number 131700250
also known as 13170
HSN 29339900
Price (INR)
13,325.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,325.00
Each
Chemical Identifiers
CAS290-37-9
IUPAC Namepyrazine
Molecular FormulaC4H4N2
InChI KeyKYQCOXFCLRTKLS-UHFFFAOYSA-N
SMILESC1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Crystals
Appearance (Color)White
Infrared spectrumConforms
Melting point52°C to 56°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only