3-Pyridinecarboxaldehyde, 98%
3-Pyridinecarboxaldehyde, 98%
3-Pyridinecarboxaldehyde, 98%
Thermo Scientific Chemicals

3-Pyridinecarboxaldehyde, 98%

CAS: 500-22-1 | C6H5NO | 107.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 131831000
Price (EUR)
75,65
Online exclusive
83,40
Save 7,75 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
75,65
Online exclusive
83,40
Save 7,75 (9%)
Each
Chemical Identifiers
CAS500-22-1
IUPAC Namepyridine-3-carbaldehyde
Molecular FormulaC6H5NO
InChI KeyQJZUKDFHGGYHMC-UHFFFAOYSA-N
SMILESO=CC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5480 to 1.5500 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only