Thermo Scientific Chemicals

2-(Hydroxymethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: 131991000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(Hydroxymethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: 131991000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
586-98-1
IUPAC Name
(pyridin-2-yl)methanol
Molecular Formula
C6H7NO
InChI Key
SHNUBALDGXWUJI-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=N1
Appearance (Color)
Clear colorless to yellow to brown
Infrared spectrum
Conforms
Refractive index
1.5410 to 1.5450 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text