Pyrrolidine, 99+%
Pyrrolidine, 99+%
Pyrrolidine, 99+%
Thermo Scientific Chemicals

Pyrrolidine, 99+%

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
数量:
100 mL
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 132080025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123-75-1
IUPAC Namepyrrolidine
Molecular FormulaC4H9N
InChI KeyRWRDLPDLKQPQOW-UHFFFAOYSA-N
SMILESC1CCNC1
さらに表示
Water=<0.2 %
Color scale=<50 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4421 to 1.4441 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only