Pyrrolidine, 99+%
Pyrrolidine, 99+%
Pyrrolidine, 99+%
Thermo Scientific Chemicals

Pyrrolidine, 99+%

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 132081000
價格 (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS123-75-1
IUPAC Namepyrrolidine
Molecular FormulaC4H9N
InChI KeyRWRDLPDLKQPQOW-UHFFFAOYSA-N
SMILESC1CCNC1
檢視更多
規格Specification Sheet規格表
Water=<0.2 %
Color scale=<50 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4421 to 1.4441 (20°C, 589 nm)
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only