2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
Thermo Scientific Chemicals

2-Pyrrolidinone, 99%

CAS: 616-45-5 | C4H7NO | 85.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 132112500
Price (EUR)
30,65
Online exclusive
33,40
Save 2,75 (8%)
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
30,65
Online exclusive
33,40
Save 2,75 (8%)
Each
Chemical Identifiers
CAS616-45-5
IUPAC Namepyrrolidin-2-one
Molecular FormulaC4H7NO
InChI KeyHNJBEVLQSNELDL-UHFFFAOYSA-N
SMILESO=C1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Infrared spectrumConforms
Appearance (Form)Liquid or low melting mass
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only