Pyruvic acid, 98%, extra pure
Pyruvic acid, 98%, extra pure
Pyruvic acid, 98%, extra pure
Thermo Scientific Chemicals

Pyruvic acid, 98%, extra pure

CAS: 127-17-3 | C3H4O3 | 88.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 132141000
Price (USD)
76.70
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.70
Each
Chemical Identifiers
CAS127-17-3
IUPAC Name2-oxopropanoic acid
Molecular FormulaC3H4O3
InChI KeyLCTONWCANYUPML-UHFFFAOYSA-N
SMILESCC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to amber
Titration with NaOH97.5 to 102.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4260 to 1.4320 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only