Quinoline, 96%
Quinoline, 96%
Quinoline, 96%
Thermo Scientific Chemicals

Quinoline, 96%

CAS: 91-22-5 | C9H7N | 129.16 g/mol
数量:
500 mL
2.5 L
10 L
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 132190025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-22-5
IUPAC Namequinoline
Molecular FormulaC9H7N
InChI KeySMWDFEZZVXVKRB-UHFFFAOYSA-N
SMILESC1=CC=C2N=CC=CC2=C1
さらに表示
Appearance (Color)Clear light yellow to dark brown
Infrared spectrumConforms
Refractive index1.6230 to 1.6270 (20°C, 589 nm)
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only