Quinoxaline, 98+%, Thermo Scientific Chemicals
Quinoxaline, 98+%, Thermo Scientific Chemicals
Quinoxaline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Quinoxaline, 98+%, Thermo Scientific Chemicals

CAS: 91-19-0 | Molecular Formula: C8H6N2 | Molecular Weight: 130.15 g/mol
Catalog NumberQuantityPackaging
132221000100 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 132221000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS91-19-0
IUPAC Namequinoxaline
Molecular FormulaC8H6N2
InChI KeyXSCHRSMBECNVNS-UHFFFAOYSA-N
SMILESC1=CC=C2N=CC=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
Melting point29°C to 34°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Low melting crystalline solid

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only