Rhodamine B, pure
Rhodamine B, pure
Rhodamine B, pure
Thermo Scientific Chemicals

Rhodamine B, pure

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 132311000
Price (USD)
61.65
Online Exclusive
68.20
Save 6.55 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.20
Save 6.55 (10%)
Each
Chemical Identifiers
CAS81-88-9
IUPAC Name9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyPYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<10 % (105°C)
Specific absorbance A (1%/1cm)>=1700 (at lambda max.)
Infrared spectrumConforms
Absorption Ratio P-15/P+151.1 to 1.6
Appearance (Form)Powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only