Rhodamine B, pure
Rhodamine B, pure
Rhodamine B, pure
Acros Organics

Rhodamine B, pure

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
132315000
also known as 13231
500 gGlass bottle
Catalog number 132315000
also known as 13231
HSN 32041329
Price (INR)
25,021.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
25,021.00
Each
Chemical Identifiers
CAS81-88-9
IUPAC Name9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyPYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Absorption Ratio P-15/P+151.1 to 1.6
Appearance (Form)Powder or crystals
UV Lambda max549 to 558 nm (in 50:50 IMS:water)
Loss on drying=<10 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only