Rutin, 97+%
Rutin, 97+%
Rutin, 97+%
Thermo Scientific Chemicals

Rutin, 97+%

CAS: 207671-50-9 | C27H30O16 | 610.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 132391000
Price (USD)
131.65
Special offer
Online exclusive
Ends: 28-Dec-2026
155.00
Save 23.35
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
131.65
Special offer
Online exclusive
Ends: 28-Dec-2026
155.00
Save 23.35
Each
Chemical Identifiers
CAS207671-50-9
IUPAC Name2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-4H-chromen-4-one
Molecular FormulaC27H30O16
InChI KeyIKGXIBQEEMLURG-NVPNHPEKSA-N
SMILESC[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Yellow to yellow-green
Titration excess NaOH97.0 to 103.0 % (On dry basis)
Ash=<0.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only