Human FGF-basic (FGF-2/bFGF) Recombinant Protein - FAQs
Human FGF-basic (FGF-2/bFGF) Recombinant Protein - FAQs
View additional product information for Human FGF-basic (FGF-2/bFGF) Recombinant Protein - FAQs (13256029)
5 product FAQs found
Can I reconstitute bFGF Recombinant Human Protein (Cat. No. 13256029) in distilled water?
As bFGF Recombinant Human Protein can lose some activity in purely aqueous solutions, we recommend resuspending the product in 10 mM Tris, pH 7.6 as recommended in the product sheet.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
It looks like the recombinant protein vial I received from you is empty. Is this normal?
Gibco recombinant proteins are frequently formulated without carrier proteins or additives (e.g., BSA, HSA, sucrose, etc.) and no Gibco PeproTech recombinant protein contains a carrier protein. As a result, during lyophilization, the protein product may be deposited on the vial as a thin, and sometimes invisible, film instead of a pellet. The size of the pellet, if any, is not directly related to the quantity of the recombinant protein in the vial. Our quality control procedures assure that each vial contains the correct amount of product.
To ensure complete recovery of protein product, before opening a vial of lyophilized recombinant protein, we recommend centrifuging it in a microcentrifuge for 20-30 seconds to drive any protein that may be lodged in the cap or on the side to the bottom of the vial. After reconstitution, you can confirm the presence of product protein by running a small amount on SDS-PAGE. In general, a protein band with expected size should be visible with as little as 10 ng of protein loaded on an acrylamide gel.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Why does my recombinant protein not show activity in my own experiment?
Assay time is critical. Each assay needs to beoptimized and performed at the peak response time. Different cells may respond differently to a growth factor or cytokine. We suggest repeating our QC assay using same indicator cells as suggested in the manual to see if you can obtain a similar response. In addition, serum may be masking the response. Serum starvation might be needed for certain types of assays.