Sesamol, 98%
Sesamol, 98%
Sesamol, 98%
Thermo Scientific Chemicals

Sesamol, 98%

CAS: 533-31-3 | C7H6O3 | 138.12 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 132690250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS533-31-3
IUPAC Name2H-1,3-benzodioxol-5-ol
Molecular FormulaC7H6O3
InChI KeyLUSZGTFNYDARNI-UHFFFAOYSA-N
SMILESOC1=CC=C2OCOC2=C1
さらに表示
Appearance (Form)Crystalline powder
Water=<0.2 % (K.F.)
Appearance (Color)Off-white to beige
AppearanceMay darken on storage
Melting point63°C to 65°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only