D-Sorbitol, 97%
D-Sorbitol, 97%
D-Sorbitol, 97%
Thermo Scientific Chemicals

D-Sorbitol, 97%

CAS: 50-70-4 | C6H14O6 | 182.17 g/mol
数量:
5 g
100 g
500 g
1 kg
2.5 kg
10 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 132730050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS50-70-4
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
さらに表示
HPLC>=96.0 %
Sulfated ash=<0.1 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Specific optical rotation(on anhydrous substance)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only