Stigmasterol, 95%, Thermo Scientific Chemicals
Stigmasterol, 95%, Thermo Scientific Chemicals
Stigmasterol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stigmasterol, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
1327600505gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 132760050
Price (USD)
-
Quantity:
5g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS83-48-7
IUPAC Name(1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,3E,5S)-5-ethyl-6-methylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC29H48O
InChI KeyHCXVJBMSMIARIN-PHZDYDNGSA-N
SMILESCC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Specific optical rotation-48° to -52° (20°C, 589 nm) (c=2, CHCl3)
Appearance (Color)White
Infrared spectrumConforms
Melting point161°C to 170°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only