Syringic acid, 97%
Syringic acid, 97%
Syringic acid, 97%
Thermo Scientific Chemicals

Syringic acid, 97%

CAS: 530-57-4 | C9H10O5 | 198.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 132890100
Price (USD)
53.60
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.60
Each
Chemical Identifiers
CAS530-57-4
IUPAC Name4-hydroxy-3,5-dimethoxybenzoic acid
Molecular FormulaC9H10O5
InChI KeyJMSVCTWVEWCHDZ-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point206°C to 212°C
Water=<0.5 % (K.F.)
Appearance (Color)Beige to grey to light brown
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only