Syringic acid, 97%
Syringic acid, 97%
Syringic acid, 97%
Thermo Scientific Chemicals

Syringic acid, 97%

CAS: 530-57-4 | C9H10O5 | 198.17 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
產品號碼 132890250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS530-57-4
IUPAC Name4-hydroxy-3,5-dimethoxybenzoic acid
Molecular FormulaC9H10O5
InChI KeyJMSVCTWVEWCHDZ-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1O)C(O)=O
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Melting point206°C to 212°C
Water=<0.5 % (K.F.)
Appearance (Color)Beige to grey to light brown
Appearance (Form)Powder
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only