1-Acetamidoadamantane, 97%
1-Acetamidoadamantane, 97%
1-Acetamidoadamantane, 97%
Thermo Scientific Chemicals

1-Acetamidoadamantane, 97%

CAS: 880-52-4 | C12H19NO | 193.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 133600250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS880-52-4
IUPAC NameN-(adamantan-1-yl)acetamide
Molecular FormulaC12H19NO
InChI KeyBCVXYGJCDZPKGV-UHFFFAOYSA-N
SMILESCC(=O)NC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
Appearance (Form)Crystalline powder or powder or crystals
Melting point145°C to 150°C
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only