Piperazine-2-carboxylic acid dihydrochloride, 98%
Piperazine-2-carboxylic acid dihydrochloride, 98%
Piperazine-2-carboxylic acid dihydrochloride, 98%
Thermo Scientific Chemicals

Piperazine-2-carboxylic acid dihydrochloride, 98%

CAS: 3022-15-9 | C5H10N2O2·2ClH | 203.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 133660500
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS3022-15-9
IUPAC Name(3R)-piperazine-1,4-diium-3-carboxylate
Molecular FormulaC5H11N2O2
InChI KeyJSSXHAMIXJGYCS-SCSAIBSYSA-O
SMILES[O-]C(=O)[C@H]1C[NH2+]CC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO497.5 to 102.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White to yellow
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only