9-Aminoacridine hemihydrate, 98%
9-Aminoacridine hemihydrate, 98%
9-Aminoacridine hemihydrate, 98%
Thermo Scientific Chemicals

9-Aminoacridine hemihydrate, 98%

CAS: 65944-23-2 | C13H10N2·1/2H2O | 203.16 g/mol
Catalog NumberQuantityPackaging
1344100101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 134410010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS65944-23-2
IUPAC Namebis(acridin-9-amine) hydrate
Molecular FormulaC26H22N4O
InChI KeySPWFQHCZHQCDAG-UHFFFAOYSA-N
SMILESO.NC1=C2C=CC=CC2=NC2=CC=CC=C12.NC1=C2C=CC=CC2=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only