Interested in a bulk or custom version of this product? Request pricing.

DCX Recombinant Human Protein, N-GST Tag

Catalog number: 13498H09E50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Doublecortin (DCX), recombinant human protein is supplied in frozen protein. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human DCX (O43602-2) N-terminal fragment (Ala 45-Val 150) fused to the GST tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 20 mM Tris, 1 mM DTT, 10% glycerol, pH 7.5.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: RP5-914P14.1, DBCN, DC, LISX, SCLH, XLIS
Gene ID: 1641
Gene Symbol: DCX
Molecular Weight: 39.4 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Cell Structure⁄Organelle Proteins
Protein Form: Recombinant, Truncated
Protein Name: Doublecortin
Protein Tag or Fusion: GST Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Neurobiology, Signal Transduction
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols