What reagents are used to coat your Nunclon Delta surface products?
The Nunclon Delta surface is a surface modification that enhances cell attachment and growth for adherent cell lines and is not coated with any chemical reagent.
Search
View additional product information for Nunc™ MiniTrays with Nunclon™ Delta surface - FAQs (136528, 163118)
1 product FAQs found