Thermal Mixers and Blocks
Actual product may vary
Thermal Mixers and Blocks
Thermal Mixers and Blocks
Thermal Mixers and Blocks
Thermal Mixers and Blocks
Thermal Mixers and Blocks
Thermo Scientific™

Thermal Mixers and Blocks

Get flexibility with this vertical mixer with variable speed and temperature control for a variety of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeHolds
13687718Mixer with Block24 x 2.0mL Microtubes
13687720Mixer with Block20 x 0.5mL, 12 x 1.5mL Microtubes
13687717Mixer with Block24 x 1.5mL Microtubes
13687719Mixer with Block20 x 0.2mL, 12 x 1.5mL Microtubes
13687721Mixer with Block96-well Microplates, 0.2mL Tube Strips
13687711Thermal MixerN/A
13687712Block24 x 1.5mL Microtubes
13687713Block24 x 2.0mL Microtubes
13687714Block20 x 2mL, 12 x 1.5mL Microtubes
13687715Block20 x 0.5mL, 12 x 1.5mL Microtubes
13687716Block96-well Microplates, 0.2mL Tube Strips
Catalog number 13687718
Price (MXN)
-
Product Type:
Mixer with Block
Holds:
24 x 2.0mL Microtubes

Gain variable speed and temperature control with the Thermo Scientific™ Thermal Mixer, which comes with a choice of blocks for microtubes and microplates.

  • Heating and cooling capability to cover a wide range of applications
  • Compact size with quiet operation
  • Convenient packages that include a Thermal Mixer with a Block

Mixer Features

  • Fast heat-up and cooling times and quick to reach specified mixing speed
  • Temperature control range : 15°C below ambient to 100°C
  • Temperature setting range : +4°C to 100°C
  • Shaking speed : 250 to 1400 rpm
  • Temperature stability : ±0.1°C
  • Even mixing across the thermoshaker block
  • Excellent uniformity across the block
  • Digital timer with sound alarm : 1 min to 96 hours
  • Temperature calibration function – compensate for differences in the thermal behavior of different tubes and samples
  • Choice of 5 interchangeable blocks
  • LCD Display indicates the set and actual time of operation, mixing speed, and temperature
  • Heating/Cooling and mixing functions can be run independently of each other
Specifications
Dimensions (L x W x H)130 x 230 x 205 mm
DisplayLCD
Electrical Requirements120/240 V
For Use With (Application)
  • Genetic analysis
  • DNA and RNA extraction
  • Sample preparation
  • Biochemical studies of enzymatic reactions and processes
  • Extraction of metabolites from cellular material
  • Incubation
  • Agitation
  • Digestion
  • Samples/standards for peptide mapping
Height (English)8.07 in.
Height (Metric)205 mm
Holds24 x 2.0mL Microtubes
Length (English)5.11 in.
Length (Metric)130 mm
Plug TypeUS, EU, UK Plug
Product TypeMixer with Block
Shipping Weight (Metric)4 kg
Timer Range1 min. to 96 hr. (1 min. increment)
Tube Capacity2.0 mL
Warranty and Service Offering2 Years
Wattage60 W
Width (English)9.05 in.
Width (Metric)230 mm
Orbit2 mm
Speed Range250 to 1400 rpm
Temperature Range (Metric)4°C to 100°C
Unit SizeEach